Simpson 260-8 12388 Black Analog Multimeter
Representative image only.
Simpson Mfr # 260-8 | 12388 – Item # EW-20006-06
$358.60USD / Each
$94.00USD / Each
for an additional
$94.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionBlack Analog Multimeter
More About this Item
  • Accuracy for the lab, rugged construction for the field
  • Excellent for adjustments such as nulling and peaking
  • Large color code scale
  • AC frequency response to 100 KHz
  • Zero RFI
  • Standard and high energy overload fusing
  • Varistor protected meter movement
  • UL 1244 listed
  • Mirrored scale
  • Broad range of accessories and probes
  • Wide range capabilities
  • Overload protection relay circuits
What's included: test leads, alligator clips, manual, batteries, fuses, and a spare 1A fuse.