Single open end serial nylon cartridge, 0.45/0.20 µm
Representative image only.
Item # EW-06479-18
A collection to fit a wide range of critical applications
  • Absolute-rated for use in many regulated processes
  • Meets a wide range of industry and government standards
PRODUCT OPTIONS
$231.00USD / Each
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaNylon
  • Micron Rating0.2
  • Filter Length (in)10
  • Filtration Area (ft²)6.9
  • Filtration Area (in²)993.6
  • Filtration Area (m²)0.641
  • DescriptionSingle open end serial nylon cartridge, 0.45/0.20 µm
Key Features
  • Absolute-rated for use in many regulated processes
  • Meets a wide range of industry and government standards
More About this Item
These SOE cartridges have a 222 O-ring configuration to create a tight seal and prevent influent bypass. They are autoclavable to 250°>F (121°>C) and in-line sterilizable to 257°>F (125°>C) for up to 30-minute cycles; tolerant to an accumulated exposure of 10 hours. The filters may also be sanitized with compatible chemical agents. All cartridges are suitable to 180°>F (82°>C) at 10 psid. Use a SOE cartridge-sump adapter kit (sold separately) to secure cartridges within housings.
The cartridges comply with US CFR Title 21 guidelines for repeated food contact, comply with USP Class VI-121°C Plastics guidelines and pass the MEM Elution Cytotoxicity Test. Aqueous extracts contain less than 0.25 EU/mL and typically exhibit low levels of non-volatile residues.
Multi-Layer Nylon Cartridges Two-pleated nylon
membranes with the top layer acting as a prefilter
to increase cartridge life and efficiency.