Snyder 6070000N45 Cone bottom tank, 110 gallons, 30° cone
Representative image only.
Snyder Mfr # 6070000N45 – Item # EW-06960-32
Tanks are ideal for small mix and batch applications
  • Available in 30 and 45 degree slopes
  • Specific gravity ratings are based on the industry's most severe calculation
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialHDLPE
  • Volume (Gal)110
  • Volume (Liters)416
  • Graduations (Gal)5
  • Graduations (Liters)17
  • Fill Port10 in
  • Max Temperature (° F)140
  • Max Temperature (° C)60
  • DescriptionCone bottom tank, 110 gallons, 30° cone
  • Warranty3 Years
3 Year Warranty
Key Features
  • Available in 30 and 45 degree slopes
  • Specific gravity ratings are based on the industry's most severe calculation
  • 3 year warranty
More About this Item
Small to medium size batch tanks save space and are perfect for smaller and short-term mixing, batch processes, and liquid storage. Cone bottom tanks are constructed high-density linear polyethylene and have a maximum storage temperature of 140°F. Specific gravity of tanks is 1.9. Material is UV stabilized for long term outdoor service. Material is FDA compliant and meets NSF standard 61. Model 06960-26 comes with a 17”W removable cover. All others come with 10”W caps. Manufactured with stratiform all thickness- walls are thicker at bottom of tank. Stands are constructed of steel and allow a nominal bottom clearance of 12” for total drain capability. Order stands seperately.