SPC UN1113 Universal Plus Spill Pads, 12" W x 13" L; 200/cs

Representative image only.
SPC Mfr # SPCUN1113 – Item # EW-86337-15
  • Chemically inert sorbent is ideal for hazardous materials clean-up

$205.00USD / Case of 200

SPECS & DESCRIPTION

Specifications & Description

  • Absorption21 gal/cs
  • Width (in)12
  • Length (in)13
  • Width (cm)27.9
  • Length (cm)33
  • DescriptionUniversal Plus Spill Pads, 12" W x 13" L; 200/cs

Key Features

  • Chemically inert sorbent is ideal for hazardous materials clean-up

More About this Item

These spill pads are designed to withstand aggressive chemical clean-up. Pads absorb 16 times their weight in chemicals, water, or oil. When incinerated, these pads produce only 0.02% ash.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-847-549-7600
Live chat