Spectra Por 131342 CE Dialysis Tubing, 25000 MWCO, 16mm/10.2mm; 33ft

Representative image only.
Spectra Por Mfr # 131342 – Item # EW-02899-46
Ideal for concentration, purification, and desalination
  • High-purity membranes with no heavy metal or sulfide contaminants
  • Low protein adsorption

PRODUCT OPTIONS

$582.00USD / Box of 1

SPECS & DESCRIPTION

Specifications & Description

  • MaterialCellulose Ester, biotech-grade
  • Max Molecular Weight Cut Off (Mwco) (Dalton)20000
  • Flat Width (mm)16
  • Diameter (mm)10
  • Length Per Pack (ft)33
  • Diameter (in)0.394
  • DescriptionCE Dialysis Tubing, 25000 MWCO, 16mm/10.2mm; 33ft

Key Features

  • High-purity membranes with no heavy metal or sulfide contaminants
  • Low protein adsorption

More About this Item

Cellulose Ester (CE) offers a wider range of more precise MWCO’s for critical separations; packaged as a 10-m roll in 0.05% sodium azide.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat