This item is now discontinued. We recommend the following as suggested replacements: EW-02899-81
Spectra Por 142935 RC Dialysis Tubing Closures, 23mm Width Max, Blue; 10/Pk
Representative image only.
Spectra Por Mfr # 142935 – Item # EW-02899-83
The most convenient method for sealing dialysis tubing without the risk of leaking
  • Easy to open and reseal
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Tubing Sealing Width (mm)23
  • ColorBlue
  • DescriptionRC Dialysis Tubing Closures, 23mm Width Max, Blue; 10/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Easy to open and reseal
  • 1 year warranty
More About this Item
Standard Closures are designed to float, providing buoyancy and vertical orientation when applied to the tubing top end. Autoclavable and available in several colors for easy sample identification.