Spectra Por G235072 Float-A-Lyzer G2 Dialysis Device, 300000 MWCO, 10 mL; 12/Pk
Representative image only.
Spectra Por Mfr # G235072 – Item # EW-02906-47
Disposable ready-to-use dialysis device eases sample loading and retrieval
  • 95 to 98% sample recovery, 99% sample purity and <10% sample dilution
  • Leak-proof screw cap allows easy pipette access for sample loading, testing, and retrieval
  • Ideal for dialyzing electro-eluted proteins and chromatography fractions
  • 1 year warranty
PRODUCT OPTIONS
$256.00USD / Pkg of 12
SPECS & DESCRIPTION
Specifications & Description
  • MaterialCellulose Ester, biotech-grade
  • Max Molecular Weight Cut Off (Mwco) (Dalton)300000
  • Min Molecular Weight Cut Off (Mwco) (Dalton)300000
  • Sample Volume (µL)10
  • Max Sample Volume (mL)10
  • ColorAmber
  • DescriptionFloat-A-Lyzer G2 Dialysis Device, 300000 MWCO, 10 mL; 12/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • 95 to 98% sample recovery, 99% sample purity and <10% sample dilution
  • Leak-proof screw cap allows easy pipette access for sample loading, testing, and retrieval
  • Ideal for dialyzing electro-eluted proteins and chromatography fractions
  • 1 year warranty
More About this Item
The Float-A-Lyzer G2 features biotech grade cellulose ester (CE) membrane. The low protein-binding, ultrapure membrane (no heavy metals and sulfides) is available in nine precise MWCO''s from 100 to 1,000,000 Daltons. The color-coded, re-sealable screw cap eliminates leaks associated with knots, closures, and needle punctures and allows repeated access for in-process testing.
The cylindrical tubing geometry prevents sample dilution and provides open access for total volume retrieval with included pipette. The included floatation ring and weighted bottom maintain vertical orientation during dialysis. This design also permits several units to fit in a single reservoir. All sizes are supplied dry with glycerol in a package of 12.