Spectra Por S/P 1 1 Dialysis Membrane Disc, 6000 to 8000 MWCO, 47 mm dia; pack of 50.
Representative image only.
Spectra Por Mfr # 132476 – Item # EW-02890-63
Available in a variety of dimensions to meet your application needs
  • Regenerated cellulose (RC) membrane for the gentle separation and purification of solutes
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialRegenerated Cellulose
  • Max Molecular Weight Cut Off (Mwco) (Dalton)8000
  • Min Molecular Weight Cut Off (Mwco) (Dalton)6000
  • Disc Diameter (mm)47
  • Description1 Dialysis Membrane Disc, 6000 to 8000 MWCO, 47 mm dia; pack of 50.
  • Warranty1 year
1 Year Warranty
Key Features
  • Regenerated cellulose (RC) membrane for the gentle separation and purification of solutes
  • 1 year warranty
More About this Item
MWCO of 6000 to 8000 Dalton