Spectra Por S/P 3 3 Membranes Flat sheets; 108 x 108 mm; 25/pack
Representative image only.
Spectra Por Mfr # 132723 – Item # EW-02900-32
Available in a variety of dimensions to meet your application needs
  • Regenerated Cellulose (RC) membrane for the gentle separation and purification of solutes
  • 6 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialRegenerated Cellulose
  • Max Molecular Weight Cut Off (Mwco) (Dalton)3500
  • Square Sheet Width (mm)108
  • Square Sheet Length (mm)108
  • Description3 Membranes Flat sheets; 108 x 108 mm; 25/pack
  • Warranty6 month
6 Month Warranty
Key Features
  • Regenerated Cellulose (RC) membrane for the gentle separation and purification of solutes
  • 6 month warranty
More About this Item
MWCO of 3500 Dalton