This item is now discontinued. We recommend the following as suggested replacements: EW-02890-64
Spectra Por S/P 4 4 RC Dialysis Membrane Disc, 12000-14000 MWCO, 33mm Dia; 50/Pk
Representative image only.
Spectra Por Mfr # 132498 – Item # EW-02890-68
Available in a variety of dimensions to meet your application needs
  • Regenerated Cellulose (RC) membrane for the gentle separation and purification of solutes
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialRegenerated Cellulose
  • Max Molecular Weight Cut Off (Mwco) (Dalton)14000
  • Min Molecular Weight Cut Off (Mwco) (Dalton)12000
  • Disc Diameter (mm)33
  • Description4 RC Dialysis Membrane Disc, 12000-14000 MWCO, 33mm Dia; 50/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Regenerated Cellulose (RC) membrane for the gentle separation and purification of solutes
  • 1 year warranty
More About this Item
MWCO of 12,000 to 14,000