Spectra Por S/P 6 6 Membranes; MWCO, 3500; flat width, 18mm; diameter, 11.5mm

Representative image only.
Spectra Por Mfr # 132590 – Item # EW-02901-30
Available in a variety of defined MWCO's for specific retentivity
  • Made of regenerated cellulose (RC) for gentle separation and purification of solutes.
  • 6 month warranty

SPECS & DESCRIPTION

Specifications & Description

  • MaterialRegenerated Cellulose
  • Max Molecular Weight Cut Off (Mwco) (Dalton)3500
  • Flat Width (mm)18
  • Diameter (mm)11.5
  • Length Per Pack (ft)50
  • Diameter (in)0.453
  • Description6 Membranes; MWCO, 3500; flat width, 18mm; diameter, 11.5mm
  • Warranty6 month
6 Month Warranty

Key Features

  • Made of regenerated cellulose (RC) for gentle separation and purification of solutes.
  • 6 month warranty

More About this Item

Prewetted in 0.05% sodium azide solution for easier use; contains traces of sulfur and heavy metals. Packaged in 50-ft lengths.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat