Spectrum Chemical Aluminum Ammonium Sulfate Dodecahydrate, Reagent ACS Grade; 4 x 2.5 kg

Representative image only.
Spectrum Chemical Mfr # A1075-2.5KG-CS4 – Item # EW-80001-98
Quality and delivery you can count on every time!
  • Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeACS Reagent
  • Empirical FormulaAlNH4(SO4)2•12H2O
  • Linear FormulaAlNH4(SO4)2•12H2O
  • ChemistryAnalytical Reagents;Solutions
  • FormCrystal
  • SynonymsAluminum Ammonium Sulfate | Ammonium Alum
  • Weight (g)10000
  • Formula Weight453.33
  • PackagingPoly Bottle
  • CAS Number7784-26-1
  • Shelf Life (days)730
  • Shelf Life (months)24
  • DescriptionAluminum Ammonium Sulfate Dodecahydrate, Reagent ACS Grade; 4 x 2.5 kg

Key Features

  • Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat