Spectrum Chemical Ascorbic Acid, Crystalline Powder, USP Grade; 100 g

Representative image only.
Spectrum Chemical Mfr # AS105-100GM – Item # EW-80023-55
Quality and delivery you can count on every time!
  • Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years

PRODUCT OPTIONS

$135.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeUSP
  • Empirical FormulaC6H8O6
  • Linear FormulaC6H8O6
  • SynonymsL-Ascorbic Acid | Vitamin C
  • FormCrystalline Powder
  • Weight (g)100
  • Formula Weight176.13
  • PackagingAmber Glass
  • Storage SymbolLIGHT SENSITIVE, OXYGEN SENSITIVE: Keep tightly closed in light-resistant containers.
  • CAS Number50-81-7
  • Shelf Life (days)730
  • Shelf Life (months)24

Key Features

  • Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat