Spectrum Chemical Iron Metal, Reduced, Powder, Reagent Grade; 12 kg

Representative image only.
Spectrum Chemical Mfr # I1035-12KG – Item # EW-80111-06
Quality and delivery you can count on every time!
  • Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years

PRODUCT OPTIONS

$1,938.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeReagent
  • Empirical FormulaFe
  • Linear FormulaFe
  • ChemistrySolutions
  • FormPowder
  • Weight (g)12000
  • Formula WeightA.W. 55.85
  • PackagingPoly Pail
  • Storage SymbolMOISTURE SENSITIVE: Keep tightly closed.
  • CAS Number7439-89-6
  • Shelf Life (days)730
  • Shelf Life (months)24
  • DescriptionIron Metal, Reduced, Powder, Reagent Grade; 12 kg

Key Features

  • Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat