Spectrum Chemical Sodium Peroxide, Reagent ACS Grade; 2.5 kg

Representative image only.
Spectrum Chemical Mfr # S1390-2.5KG – Item # EW-80193-04
Quality and delivery you can count on every time!
  • Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeACS Reagent
  • Empirical FormulaNa2O2
  • Linear FormulaNa2O2
  • ClassUN1504, Sodium peroxide, 5.1, PG I
  • ChemistryAnalytical Reagents;Solutions
  • Weight (g)2500
  • Formula Weight77.98
  • PackagingSteel Can
  • Storage SymbolMOISTURE SENSITIVE; AIR SENSITIVE: Keep tightly closed.
  • CAS Number1313-60-6
  • UN NumberUN1504
  • Shelf Life (days)730
  • Shelf Life (months)24
  • DescriptionSodium Peroxide, Reagent ACS Grade; 2.5 kg

Key Features

  • Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat