Spectrum Chemical Water, HPLC Grade; 1 L

Representative image only.
Spectrum Chemical Mfr # HP902-1LT – Item # EW-80109-67
Quality and delivery you can count on every time!
  • Water meets ACS Reagent grade standards

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeHPLC
  • Empirical FormulaH2O
  • Linear FormulaH2O
  • ChemistrySolutions
  • SynonymsDeionized Water
  • Volume (mL)1000
  • Formula Weight18.02
  • PackagingAmber Glass
  • CAS Number7732-18-5
  • Shelf Life (days)730
  • Shelf Life (months)24
  • DescriptionWater, HPLC Grade; 1 L

Key Features

  • Water meets ACS Reagent grade standards

More About this Item

Water, Reagent, ACS is used in many chemical reactions. Due to its ability to dissolve many ionic compounds, water is a common solvent in inorganic reactions. It is, however, rarely used as a reaction solvent since it is amphoteric and is not able to dissolve reactants very well. As an ACS grade Reagent, Spectrum Chemical manufactured water is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat