Spectrum Chemical Water, HPLC Grade, 6 x 1 L

Representative image only.
Spectrum Chemical Mfr # HP902-1LT-CS6 – Item # EW-80109-68
Quality and consistency your lab can count on
  • Water meets HPLC grade standards
  • Water comes in six 1 L amber glass bottles

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeHPLC
  • Empirical FormulaH2O
  • FormLiquid
  • Synonymsdihydrogen oxide, dihydrogen monoxide
  • Volume (mL)6000
  • Formula Weight18.02
  • PackagingAmber Glass
  • DOT InformationNot regulated by DOT
  • CAS Number7732-18-5
  • Shelf Life (months)24
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • DescriptionWater, HPLC Grade, 6 x 1 L

Key Features

  • Water meets HPLC grade standards
  • Water comes in six 1 L amber glass bottles

More About this Item

Water, HPLC Grade, is used in most HPLC mobile phases and in sample preparation. Because it dissolves most substances and is nontoxic, it is known as a Universal Solvent. To be used for HPLC analysis, it must conform to the highest purity standards. All HPLC solvents offered by Spectrum are glass distilled, sub-micron filtered, and undergo rigorous specification testing to insure accurate, repeatable results.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat