Spex CertiPrep 2-CLVIN 2-Chloroethyl Vinyl Ether Organic Standard, 2,000 µg/mL; 1 mL

Representative image only.
Spex CertiPrep Item # EW-04566-21
Analytical standard for monitoring purgeable VOCs in finished drinking water
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Methods 524, 624, and 8260
  • 2,000 ppm 2-chloroethyl vinyl ether organic standard in methanol-P&T matrix
  • 1 mL ampule

$32.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Concentration2,000 µg/mL (2,000 ppm)
  • MatrixMethanol-P&T
  • Element/Ion2-Chloroethyl vinyl ether
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Methanol; Class 3, 6.1, Packing Group II
  • Storage SymbolRefrigerator
  • Hazard SymbolFlammable, Acute Toxicity, Health Hazard
  • CAS Number110-75-8
  • UN NumberUN1230
  • Shelf Life (months)24
  • Description2-Chloroethyl Vinyl Ether Organic Standard, 2,000 µg/mL in Methanol-P&T; 1 mL

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Methods 524, 624, and 8260
  • 2,000 ppm 2-chloroethyl vinyl ether organic standard in methanol-P&T matrix
  • 1 mL ampule

More About this Item

Applicable Methods:
• US EPA Method 524 is a GC/MS method for the analysis of purgeable VOCs in finished drinking water. Analytes added to Methanol 524.3 Revision 4 are supplied in a 24-component mix.
• US EPA Method 624 is a general analytical method for the monitoring of VOCs in municipal and industrial wastewater by GC/MS.
• US EPA Method 8260 is a general analytical method for the monitoring of VOCs by GC/MS.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat