Spex CertiPrep 605-X Benzidines Mix for EPA Method 605, 8270 & CLP Series Methods, 2,000 µg/mL; 1 mL

Representative image only.
Spex CertiPrep Item # EW-04557-09
Analytical standard for the analysis of industrial and environmental wastewater
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Certified by DQS to ISO 9001:2015
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 2,000 ppm benzidine mix in methanol matrix
  • 1 mL ampule

SPECS & DESCRIPTION

Specifications & Description

  • Concentration2,000 µg/mL (2,000 ppm)
  • MatrixMethanol
  • Element/IonBenzidine and 3,3’-Dichlorobenzidine
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Methanol; Class 3, 6.1, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Acute Toxicity, Irritant, Health Hazard
  • CAS Number92-87-5, 91-94-1
  • UN NumberUN1230
  • Shelf Life (months)36
  • DescriptionBenzidines Mix for US EPA Methods 605, 8270 & CLP Series Methods, 2,000 µg/mL; 1 mL

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Certified by DQS to ISO 9001:2015
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 2,000 ppm benzidine mix in methanol matrix
  • 1 mL ampule

More About this Item

US EPA Method 605 is an analytical method for the monitoring of benzidine in municipal and industrial wastewater by HPLC and an Electrochemical Detector.

US EPA Method 8270 is a general analytical method for the monitoring of semivolatiles in environmental and waste samples by GC/MS.

CLP Series - The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP Semivolatile Organic Analysis Method is an analytical method for the analysis of aqueous or solid samples for SVOCs by GC/MS.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat