Spex CertiPrep 8260-BIG-MIX Volatile Organics Mix (76 Certified Components), 2,000 µg/mL; 1 mL

Representative image only.
Spex CertiPrep Item # EW-04556-92
Analytical standards for drinking water, wastewater, and solid waste
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Method 8260 and CLP Series Methods
  • 2,000 ppm volatile organic mix (76 certified components) in methanol-P&T matrix
  • 1 mL ampule

SPECS & DESCRIPTION

Specifications & Description

  • Concentration2,000 µg/mL (2,000 ppm)
  • MatrixMethanol-P&T
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Methanol; Class 3, Packing Group II
  • Storage SymbolFreezer
  • Hazard SymbolFlammable, Acute Toxicity, Irritant, Health Hazard
  • CAS Number75-05-8, 107-13-1, 107-05-1, 71-43-2, 108-86-1, 74-97-5, 75-27-4, 75-25-2, 104-51-8, 135-98-8, 98-06-6, 75-15-0, 56-23-5, 126-99-8, 108-90-7, 107-07-3, 67-66-3, 95-49-8, 106-43-4, 96-12-8, 124-48-1, 106-93-4, 74-95-3, 95-50-1, 541-73-1, 106-46-7, 1476-11-5, 110-57-6, 75-34-3, 107-06-2, 75-35-4, 156-59-2, 156-60-5, 78-87-5, 142-28-9, 594-20-7, 563-58-6, 10061-01-5, 10061-02-6, 123-91-1, 60-29-7, 97-63-2, 100-41-4, 87-68-3, 74-88-4, 98-82-8, 99-87-6, 126-98-7, 96-33-3, 80-62-6, 75-09-2, 78-83-1, 91-20-3, 98-95-3, 79-46-9, 76-01-7, 107-12-0, 103-65-1, 100-42-5, 630-20-6, 79-34-5, 127-18-4, 109-99-9, 108-88-3, 87-61-6, 120-82-1, 71-55-6, 79-00-5, 79-01-6, 96-18-4, 76-13-1, 95-63-6, 108-67-8, 108-38-3, 95-47-6, 106-42-3
  • UN NumberUN1230
  • Shelf Life (months)12
  • DescriptionVolatile Organics Mix (76 Certified Components), 2,000 µg/mL in Methanol-P&T; 1 mL

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Method 8260 and CLP Series Methods
  • 2,000 ppm volatile organic mix (76 certified components) in methanol-P&T matrix
  • 1 mL ampule

More About this Item

US EPA Method 8260 is a general analytical method for the monitoring of VOCs by GC/MS.

CLP Series Methods - The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP Volatile Organic Analysis Method is an analytical method for the analysis of aqueous or solid samples for VOCs by GC/MS.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-component solution contains 76 components.

• Volatile Organics Mix is a 1 mL ampule which contains 2,000 µg/mL (2,000 ppm) of Acetonitrile, Acrylonitrile, Allyl chloride, Benzene, Bromobenzene, Bromochloromethane, Bromodichloromethane, Bromoform, n-Butylbenzene, sec-Butylbenzene, tert-Butylbenzene, Carbon disulfide, Carbon tetrachloride, 2-Chloro-1,3-butadiene, Chlorobenzene, 2-Chloroethanol, Chloroform, 2-Chlorotoluene, 4-Chlorotoluene, 1,2-Dibromo-3-chloropropane, Dibromochloromethane, 1,2-Dibromoethane, Dibromomethane, 1,2-Dichlorobenzene, 1,3-Dichlorobenzene, 1,4-Dichlorobenzene, cis-1,4-Dichloro-2-butene, trans-1,4-Dichloro-2-butene, 1,1-Dichloroethane, 1,2-Dichloroethane, 1,1-Dichloroethene, cis-1,2-Dichloroethene, trans-1,2-Dichloroethene, 1,2-Dichloropropane, 1,3-Dichloropropane, 2,2-Dichloropropane, 1,1-Dichloropropene, cis-1,3-Dichloropropene, trans-1,3-Dichloropropene, 1,4-Dioxane, Ether, Ethyl methacrylate, Ethylbenzene, Hexachlorobutadiene, Iodomethane, Isopropylbenzene, p-Isopropyltoluene, Methacrylonitrile, Methyl acrylate, Methyl methacrylate, Methylene chloride, 2-Methyl-1-propanol, Naphthalene, Nitrobenzene, 2-Nitropropane, Pentachloroethane, Propionitrile, n-Propylbenzene, Styrene, 1,1,1,2-Tetrachloroethane, 1,1,2,2-Tetrachloroethane, Tetrachloroethene, Tetrahydrofuran, Toluene, 1,2,3-Trichlorobenzene, 1,2,4-Trichlorobenzene, 1,1,1,-Trichloroethane, 1,1,2-Trichloroethane, Trichloroethene, 1,2,3-Trichloropropane, 1,1,2-Trichlorotrifluoroethane, 1,2,4-Trimethylbenzene, 1,3,5-Trimethylbenzene, m-Xylene, o-Xylene, and p-Xylene.

Hazardous material shipping fees may apply.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat