Spex CertiPrep 930 Uranium Drinking Water QC Sample, Ready-to-Analyze; 500 mL

Representative image only.
Spex CertiPrep Item # EW-04573-40
Meet or exceed performance criteria with QC samples
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for AA, ICP, ICP-MS, IC, XRF, and other analytical instrumentation
  • For ASTM D5673-03, US EPA Method 200.8, and SM 18/19th ED 3125
  • For corrosivity calcium carbonate saturation and Langelier saturation index
  • Ready-to-analyze 500 mL bottle

$159.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Concentration3 to 104 µg/L
  • ClassQC Samples
  • Element/IonUranium (Natural)
  • FormLiquid
  • Volume (mL)500
  • DOT InformationNon-Hazardous
  • Storage SymbolAmbient
  • DescriptionUranium Drinking Water QC Sample, Ready-to-Analyze; 500 mL

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for AA, ICP, ICP-MS, IC, XRF, and other analytical instrumentation
  • For ASTM D5673-03, US EPA Method 200.8, and SM 18/19th ED 3125
  • For corrosivity calcium carbonate saturation and Langelier saturation index
  • Ready-to-analyze 500 mL bottle

More About this Item

The end-product of most analytical laboratories is data. It is therefore imperative that the quality of the data produced meet or exceed performance criteria. The implementation of Quality Control (QC) processes helps control error and estimate uncertainty. QC can be a tool in method development, regulatory processes, laboratory training, and performance monitoring. QC samples can help an analytical laboratory determine its precision, accuracy, linear range, and method ruggedness.

QC samples allow the laboratories to fulfill documentation requirements as well as correct errors in root case analysis of a possible failure. Finally, QC samples monitor the effectiveness of the laboratory analysts to produce accurate data.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat