Spex CertiPrep AS-CL9-2Y Chloride Single-Element Ion Anion Standard, 1,000 µg/mL in H2O; 125 mL

Representative image only.
Spex CertiPrep Item # EW-04501-31
Single element standard for ion chromatography
  • Ideal for a variety of different kinds of charged analytes
  • Manufactured under ISO ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
  • Contains 1,000 ppm of Chloride (CI)- in H20 matrix

PRODUCT OPTIONS

$77.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixH2O
  • ClassIC
  • Element/IonChloride
  • FormLiquid
  • Volume (mL)125
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • Hazard SymbolNot regulated
  • UN NumberNot regulated
  • Shelf Life (days)365
  • Shelf Life (months)12
  • DescriptionChloride Single-Element Ion Anion Standard, 1,000 µg/mL in H2O; 125 mL

Key Features

  • Ideal for a variety of different kinds of charged analytes
  • Manufactured under ISO ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
  • Contains 1,000 ppm of Chloride (CI)- in H20 matrix

More About this Item

Ion chromatography (IC) is an analytical process for the separation of ions based on charge affinity. IC can be used for a variety of different kinds of charged analytes from single elements to large proteins. In order to ensure accurate analysis, quality standards which are traceable and stable are necessary.

These solutions offer the highest quality IC standards available for the analytical laboratory. Spex CertiPrep®Certified Reference Materials (CRMs) are manufactured under a quality system complying with the requirements of ISO 9001, ISO/IEC 17025 and ISO 17034.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat