Spex CertiPrep AS-SO49-2X Sulfate (SO4)-2 Single-Element Ion Anion Standard, 1,000 µg/mL (1,000 ppm) in H2O, 500 mL

Representative image only.
Spex CertiPrep Item # EW-04501-82
Single and multi-element standards for ion chromatography
  • Traceable standards
  • Perfect for the analytical laboratory

PRODUCT OPTIONS

$141.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixWater
  • Element/IonSulfate
  • FormLiquid
  • Volume (mL)500
  • Storage SymbolAmbient
  • Shelf Life (days)365
  • Shelf Life (months)12
  • DescriptionSulfate (SO4)-2 Single-Element Ion Anion Standard, 1,000 µg/mL (1,000 ppm) in H2O, 500 mL

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Traceable standards
  • Perfect for the analytical laboratory

More About this Item

Ion chromatography (IC) is an analytical process for the separation of ions based on charge affinity. IC can be used for a variety of different kinds of charged analytes from single elements to large proteins. To ensure accurate analysis, quality standards which are traceable and stable are necessary. Spex CertiPrep®offers the highest quality IC standards available for the analytical laboratory.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at CRMsales@spex.com or call us at 732-549-7144.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat