Spex CertiPrep CLP90-X Carbazole Stock Solution for CLP Series Methods, 2,000 µg/mL, 1 mL

Representative image only.
Spex CertiPrep Item # EW-04566-00
Analytical standard for analysis of aqueous or solid samples for VOCs
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For CLP Series Methods
  • 2,000 ppm carbazole stock solution in methylene chloride matrix
  • 1 mL ampule

Call For Price

SPECS & DESCRIPTION

Specifications & Description

  • Concentration2,000 µg/mL (2,000 ppm)
  • MatrixMethylene Chloride
  • Element/IonCarbazole
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Dichloromethane; Class 6.1, Packing Group III
  • Storage SymbolFreezer
  • Hazard SymbolIrritant, Health Hazard
  • UN NumberUN1593
  • CAS Number86-74-8
  • Shelf Life (months)36
  • DescriptionCarbazole Stock Solution for CLP Series Methods, 2,000 µg/mL in Methylene Chloride, 1 mL

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For CLP Series Methods
  • 2,000 ppm carbazole stock solution in methylene chloride matrix
  • 1 mL ampule

More About this Item

Applicable Methods:
• CLP Series Methods: The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP Volatile Organic Analysis Method is an analytical method for the analysis of aqueous or solid samples for VOCs by GC/MS.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat