Spex CertiPrep CLPS-MSB-TI Base/Neutrals Tinted Matrix Spike for EPA & CLP, 1,000 µg/mL; 1mL

Representative image only.
Spex CertiPrep Item # EW-04557-85
Analytical standard for monitoring municipal water, industrial wastewater, and environmental and waste samples
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Methods 625, 8270 and CLP Series Methods
  • 1,000 ppm base/neutrals tinted matrix spike in methanol matrix
  • 1 mL ampule

$49.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixMethanol
  • Element/Ion1,2,4-Trichlorobenzene, 1,4-Dichlorobenzene, 2,4-Dinitrotoluene, Acenaphthene, Bromophenol blue, n-Nitrosodi-n-propylamine, Pyrene
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Methanol; Class 3, 6.1, Packing Group II
  • Storage SymbolFreezer
  • Hazard SymbolFlammable, Acute Toxicity, Health Hazard
  • CAS Number120-82-1, 106-46-7, 121-14-2, 83-32-9, 115-39-9, 621-64-7, 129-00-0
  • UN NumberUN1230
  • Shelf Life (months)36
  • DescriptionBase/Neutrals Tinted Matrix Spike for US EPA Methods 625, 8270 and CLP Series Methods, 1,000 µg/mL in Methanol; 1mL

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Methods 625, 8270 and CLP Series Methods
  • 1,000 ppm base/neutrals tinted matrix spike in methanol matrix
  • 1 mL ampule

More About this Item

US EPA Method 625 is an analytical method for the monitoring of acid and base neutral extractable fractions in municipal and industrial wastewater by GC/MS.

US EPA Method 8270 is a general analytical method for the monitoring of semivolatiles in environmental and waste samples by GC/MS.

CLP Series Methods: The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP Volatile Organic Analysis Method is an analytical method for the analysis of aqueous or solid samples for VOCs by GC/MS.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat