Spex CertiPrep FFB-100-03 Lithium Bromide Crystals Ultra Pure Additive; 125 g

Representative image only.
Spex CertiPrep Item # EW-04570-92
Additives for XRF, ICP and AA
  • Prevent borate glass from sticking to the platinum ware
  • Ultra pure grade—99.998% purity
  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
  • Lithium bromide crystals
  • 125 g bottle

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeUltra pure
  • FormSolid
  • Weight (g)125
  • DOT InformationHazardous; Toxic solid, inorganic, n.o.s. (lithium bromide); Class 6.1, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolAcute toxicity
  • UN NumberUN3288
  • Shelf Life (months)12
  • DescriptionLithium Bromide Crystals Ultra Pure Additive; 125 g

Key Features

  • Prevent borate glass from sticking to the platinum ware
  • Ultra pure grade—99.998% purity
  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
  • Lithium bromide crystals
  • 125 g bottle

More About this Item

Nonwetting additives act as a releasing agent to reduce the risk of sample disk sticking to the platimum ware. This makes it easier to remove the disk after fusion, protecting your sample and equipment.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Hazardous material shipping fees may apply.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat