Spex CertiPrep FLAVIN-1 Flavonoid Organic Standard in Dimentyl Sulfoxide, 1,000 µg/mL, 1 mL

Representative image only.
Spex CertiPrep Item # EW-41013-17
Single-component organic standards ensure consistent results
  • Manufactured under ISO 17034 compliance

$373.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixDimethyl Sulfoxide
  • Element/IonFlavonoid
  • FormLiquid
  • Volume (mL)1
  • Storage SymbolFreezer
  • DescriptionFlavonoid Organic Standard in Dimentyl Sulfoxide, 1,000 µg/mL, 1 mL

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Manufactured under ISO 17034 compliance

More About this Item

Manufactured in an ISO 17034 facility, this certified reference standard for flavonoid analysis includes the most common flavonoids in one mix for easy use, and error and contamination reduction.

Flavonoids are naturally occurring secondary metabolic products which can have important functions within plants and benefit consumers with health and healing properties.

Many beneficial compounds are metabolites produced as an end product of chemical and biological processes. Metabolites are small molecules that have many functions including defense, pigments, pheromones, odorants, and catalysts. Primary metabolites are necessary for growth, development, and reproduction. Flavonoids are secondary plant, algae or fungus metabolites composed of polyphenolic compounds. Secondary metabolites are not directly involved in critical processes but have secondary functions involving defense and pigmentation.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at CRMsales@spex.com or call us at 732-549-7144.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat