Spex CertiPrep Hydroxylamine HCl, 6% w/v; 10 L

Representative image only.
Spex CertiPrep Item # EW-84410-32
Manufactured using the highest quality raw materials to ensure low-level impurities and consistent lot-to-lot quality
  • Designed to work with any instrument and application
  • 10 L Cubitainer® container

$1,010.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Concentration6% w/v
  • ChemistrySolvents
  • Element/IonHydroxylamine HCl
  • FormLiquid
  • Volume (mL)10000
  • PackagingCubitainer® container
  • DOT InformationNonhazardous; Not Regulated
  • Storage SymbolAmbient
  • Hazard SymbolIrritant
  • CAS Number5470-11-1
  • UN NumberNot Regulated
  • Shelf Life (months)12
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • Melting Point C (° C)-6
  • Melting Point F (° F)21.2
  • DescriptionHydroxylamine HCl, 6% w/v; 10 L

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Designed to work with any instrument and application
  • 10 L Cubitainer® container

More About this Item

Density: 1.0259
Solubility: miscible
Color: colorless
Odor: odorless

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat