Spex CertiPrep Iodine, 0.1 N, Standardized; 4 L

Representative image only.
Spex CertiPrep Item # UX-84410-35
Manufactured using the highest quality raw materials to ensure low-level impurities and consistent lot-to-lot quality
  • Designed to work with any instrument and application
  • 4 L safety-coated amber glass bottle

$190.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Concentration0.1 N
  • ChemistrySolvents
  • Element/IonIodine
  • FormLiquid
  • Volume (mL)4000
  • PackagingAmber glass bottle
  • DOT InformationNonhazardous; Not Regulated
  • Storage SymbolAmbient
  • Hazard SymbolHealth Hazard
  • UN NumberNot Regulated
  • CAS Number7553-56-2
  • Shelf Life (months)12
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • Melting Point C (° C)0
  • Melting Point F (° F)32
  • DescriptionIodine, 0.1 N, Standardized; 4 L

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Designed to work with any instrument and application
  • 4 L safety-coated amber glass bottle

More About this Item

Density: 1.05
Composition: water 96.29%, potassium Iodide 2.47%, iodine 1.24%
Solubility: miscible
Color: dark brown liquid

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat