Spex CertiPrep Iodine Monochloride Solution WIJS; 4 L

Representative image only.
Spex CertiPrep Item # EW-84410-34
Manufactured using the highest quality raw materials to ensure low-level impurities and consistent lot-to-lot quality
  • Designed to work with any instrument and application
  • 4 L amber glass bottle

$575.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • ChemistrySolvents
  • Element/IonIodine Monochloride
  • FormLiquid
  • Volume (mL)4000
  • PackagingAmber glass bottle, safety coated
  • DOT InformationHazardous; CORROSIVE LIQUIDS, FLAMMABLE, N.O.S. (ACETIC ACID, IODINE MONOCHLORIDE); Class 8 (3), Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Corrosive
  • CAS Number7553-56-2
  • UN NumberUN2920
  • Boiling Point C (° C)118
  • Boiling Point F (° F)244.4
  • Melting Point C (° C)16
  • Melting Point F (° F)60.8
  • DescriptionIodine Monochloride Solution WIJS; 4 L

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Designed to work with any instrument and application
  • 4 L amber glass bottle

More About this Item

Auto-ignition temperature: 463 °C (acetic acid)
Flash point: 39°C
Evaporation rate: 0.97 (butyl acetate = 1)
Relative density: 1.05
Vapor density: 2.1
Vapor pressure: 11.4 mmHg at 20°C
Solubility: miscible
pH range: < 2
Color: brown
Odor: Vinegar
Odor threshold: 0.074 ppm
Flammability/Explosive limits: 4 to 19.9% (Acetic Acid)
Partition coeffieient (n-octanol/water): -0.31 at 20°C

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous shipping fee may apply.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat