Spex CertiPrep LCS-5782 Imiprothrin LC/MS Standard, 1,000 µg/mL in HPLC Acetone, 1 mL

Representative image only.
Spex CertiPrep Item # EW-04562-75
LC/MS single-component organic standards reduce potential interference and increase analytical accuracy
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 1,000 ppm imiprothrin LC/MS organic standard in HPLC acetone matrix, 1 mL

$274.05USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixHPLC Acetone
  • ChemistryStandards;Organics
  • Element/IonImiprothrin
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Acetone; Class 3, Packing Group II
  • Storage SymbolFreezer
  • Hazard SymbolFlammable, Irritant
  • CAS Number72963-72-5
  • UN NumberUN1090
  • Shelf Life (months)12
  • DescriptionImiprothrin LC/MS Organic Standard, 1,000 µg/mL in HPLC Acetone, 1 mL

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 1,000 ppm imiprothrin LC/MS organic standard in HPLC acetone matrix, 1 mL

More About this Item

Spex CertiPrep® creates single and multi-component standards suitable for all HPLC detectors. Our standards are created in HPLC and LC/MS grade solvents to reduce potential interference and increase analytical accuracy.

High Performance Liquid Chromatography (HPLC) is a column-based chromatographic technique used to separate components in a mixture for identification and quantification. A pump system passes pressurized mobile phase and sample through a pre-filled column containing a packing or sorbent which aids in the separation of component peaks. As each analyte in the mixture interacts with the column material, it retards the flow of the analytes within the column. HPLC can be used for identification and quantification of compounds in a variety of fields from agriculture to medical and manufacturing.

A minimal HPLC instrument configuration includes a sampler, pumps, column compartment, and a detector. There are a multitude of various detectors commonly in use with a HPLC system including: Refractive Index Detector (RID), UV/Vis Detector, Photoiode Array Detector, and Mass Spectrometers. Each of these detectors have their own range of use and sensitivity, from the broad range RID detector with its higher detection limits, to the powerful and sensitive LC/MS techniques used for extremely low levels of detection.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat