Spex CertiPrep m-Cresol; 4 L

Representative image only.
Spex CertiPrep Item # EW-84410-42
Manufactured using the highest quality raw materials to ensure low-level impurities and consistent lot-to-lot quality
  • Designed to work with any instrument and application
  • 4 L amber glass bottle

$1,145.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Linear FormulaC7H8O
  • ChemistrySolvents
  • Element/Ionm-Cresol
  • FormLiquid
  • SynonymsMeta-cresol
  • Volume (mL)4000
  • PackagingAmber glass bottle
  • DOT InformationHazardous; Cresols, Liquid; Class 6.1 (8), Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolCorrosive, Acute Toxicity
  • CAS Number108-39-4
  • UN NumberUN2076
  • Shelf Life (months)18
  • Boiling Point C (° C)203
  • Boiling Point F (° F)397.4
  • Descriptionm-Cresol; 4 L

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Designed to work with any instrument and application
  • 4 L amber glass bottle

More About this Item

Melting point: 8 to 10 °C
Flash point: 86 °C
Density: 1.03
Molecular weight: 108.14 g/mol
Chemical formula: C7H8O
Composition: m-Cresol 100%
Solubility: 24 g/L at 25°C
Refractive index: 1.539 to 1.541nD
Color: colorless to slightly yellow liquid

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous shipping fee may apply.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat