Spex CertiPrep PH-BUFF8-500 pH 8 Certified Buffer Standard, 8 SI Units in H2O, 500 mL
Representative image only.
Spex CertiPrep Item # EW-04569-78
Premium grade for calibration and quality control checks
  • Analyzed for certified value and homogeneity
  • Certified to ISO/IEC 17025:2005 and ISO 17034:2016
PRODUCT OPTIONS
$41.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MatrixH2O
  • Element/Ion8 SI Units
  • Volume (mL)500
  • Storage SymbolAmbient
  • Shelf Life (days)365
  • Shelf Life (months)12
  • DescriptionpH 8 Certified Buffer Standard, 8 SI Units in H2O, 500 mL
Includes
Supplier Certificate of Analysis (CoA) is included with product shipment.
Key Features
  • Analyzed for certified value and homogeneity
  • Certified to ISO/IEC 17025:2005 and ISO 17034:2016
More About this Item
Spex CertiPrep® pH buffers are accurate and certified to ISO/IEC 17025:2005 and ISO 17034:2016 requirements. Made using high purity starting materials and ASTM Type I water, they have been thoroughly analyzed for certified value and homogeneity.