Spex CertiPrep Pyridine, ACS Grade; 4 L

Representative image only.
Spex CertiPrep Item # EW-84410-67
Manufactured using the highest quality raw materials to ensure low-level impurities and consistent lot-to-lot quality
  • Designed to work with any instrument and application
  • ACS grade—meets or exceeds purity standards set by the American Chemical Society
  • 4 L safety-coated amber glass bottle

$501.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeACS
  • Linear FormulaC5H5N
  • ChemistrySolvents
  • Element/IonPyridine ACS
  • SynonymsAzoBenzene, azine
  • FormLiquid
  • Volume (mL)4000
  • PackagingAmber glass bottle, safety coated
  • DOT InformationHazardous; Pyridine; Class 3, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Irritant, Health Hazard
  • CAS Number110-86-1
  • UN NumberUN1282
  • Shelf Life (months)60
  • Melting Point C (° C)-43.6
  • Melting Point F (° F)-42
  • Refractive Index1.509
  • DescriptionPyridine, ACS Grade; 4 L

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Designed to work with any instrument and application
  • ACS grade—meets or exceeds purity standards set by the American Chemical Society
  • 4 L safety-coated amber glass bottle

More About this Item

Boiling point: 115 to 116°C
Flash point: 17 °C
Density: 0.98
Molecular weight: 79.1 g/mol
Chemical formula: C5H5N
Composition: pyridine 100%
Solubility: miscible
pH range: 8.5 to 8.9
Color: colorless liquid

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous shipping fee may apply.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat