Spex CertiPrep S-2585B Isopropanol (2-propanol,isopropyl alcohol), 1,000 µg/mL; 1 mL

Representative image only.
Spex CertiPrep Item # EW-04549-82
Single-component organic standards ensure consistent results
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 1,000 ppm isopropanol (2-propanol,isopropyl alcohol) single-component organic standard in methanol matrix
  • 1 mL ampule

$80.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixMethanol
  • ChemistryOrganics;Standards
  • Element/IonIsopropanol (2-propanol,isopropyl alcohol)
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Methanol; Class 3, 6.1, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Acute Toxicity, Health Hazard
  • UN NumberUN1230
  • CAS Number67-63-0
  • Shelf Life (months)36
  • Boiling Point C (° C)97
  • Boiling Point F (° F)206.6
  • Melting Point C (° C)-127
  • Melting Point F (° F)-196.6
  • Polarity Index4
  • Solubility in Water (% wt/wt)100
  • Specific Gravity0.804
  • UV Cutoff (nm)210
  • DescriptionIsopropanol (2-propanol,isopropyl alcohol) Organic Standard, 1,000 µg/mL; 1 mL

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 1,000 ppm isopropanol (2-propanol,isopropyl alcohol) single-component organic standard in methanol matrix
  • 1 mL ampule

More About this Item

Single-component standards offer high-quality, consistent results while saving preparation time. Isopropanol (2-propanol,isopropyl alcohol) standard is supplied in methanol. Additional solvents include methylene chloride, acetone, benzene, hexane, acetonitrile, THF, and water. Please inquire about the matrix for a particular item when ordering, if the matrix is important to your application, a substitution may be possible.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at CRMsales@spex.com or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat