Spex CertiPrep S-3291 Sodium Fluoroacetate, 1,000 µg/mL in DI Water; 1 mL

Representative image only.
Spex CertiPrep Item # EW-04554-17
Single-component organic standards ensure consistent results
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 1,000 ppm sodium fluoroacetate in DI water matrix

$88.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixDI Water
  • Element/IonSodium fluoroacetate
  • FormLiquid
  • Volume (mL)1
  • DOT InformationNot Regulated
  • Storage SymbolRefrigerator
  • Hazard SymbolNot Regulated
  • CAS Number62-74-8
  • UN NumberNot Regulated
  • Shelf Life (months)36
  • DescriptionSodium Fluoroacetate, 1,000 µg/mL in DI Water; 1 mL

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 1,000 ppm sodium fluoroacetate in DI water matrix

More About this Item

Single-component standards offer high-quality, consistent results while saving preparation time. Sodium fluoroacetate standard is supplied in DI water. Additional solvents include acetone, methanol, methylene chloride, hexane, benzene, acetonitrile, and THF. Please inquire about the matrix for a particular item when ordering, if the matrix is important to your application, a substitution may be possible.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat