Spex CertiPrep S-795 Chloroacetic Acid, 1,000µg/mL in Methyl Tertiary-Butyl Ether; 1mL

Representative image only.
Spex CertiPrep Item # EW-04544-62
Single-component organic standards ensure consistent results
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 1,000 ppm chloroacetic acid single-component organic standard in methyl tertiary-butyl ether matrix
  • 1 mL ampule

$45.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixMethyl Tertiary-Butyl Ether
  • Element/IonChloroacetic acid
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Methyl tert-butyl ether; Class 3, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Acute Toxicity, Irritant
  • UN NumberUN2398
  • CAS Number79-11-8
  • Shelf Life (months)36
  • DescriptionChloroacetic Acid Organic Standard, 1,000µg/mL in Methyl Tertiary-Butyl Ether; 1 mL

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 1,000 ppm chloroacetic acid single-component organic standard in methyl tertiary-butyl ether matrix
  • 1 mL ampule

More About this Item

Single-component standards offer high-quality, consistent results while saving preparation time. Chloroacetic acid standard is supplied in methyl tertiary-butyl ether. Additional solvents include methanol, methylene chloride, acetone, benzene, hexane, acetonitrile, THF, and water. Please inquire about the matrix for a particular item when ordering, if the matrix is important to your application, a substitution may be possible.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat