Spex CertiPrep Sodium Standard, 100 ppm; 500 mL

Representative image only.
Spex CertiPrep Item # EW-84410-84
Manufactured using the highest quality raw materials to ensure low-level impurities and consistent lot-to-lot quality
  • Designed to work with any instrument and application
  • 500 mL natural poly bottle

$47.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Concentration100 µm (100 ppm)
  • ChemistrySolvents
  • Element/IonSodium
  • SynonymsSodium ion standard
  • FormLiquid
  • Volume (mL)500
  • PackagingNatural poly bottle
  • DOT InformationNonhazardous; Not Regulated
  • Storage SymbolAmbient
  • Hazard SymbolNot Regulated
  • CAS Number7647-14-5
  • UN NumberNot Regulated
  • Shelf Life (months)12
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • Melting Point C (° C)0
  • Melting Point F (° F)32
  • DescriptionSodium Standard, 100 ppm; 500 mL

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Designed to work with any instrument and application
  • 500 mL natural poly bottle

More About this Item

Density: 1
Composition: water 99.97%, sodium chloride 0.03%
Solubility: miscible
Color: colorless liquid

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat