Spex NanoSNAP™ Maintenance Kit, Hydrophobic Reagent and Swabs
Representative image only.
Spex Item # EW-83057-03
Optimize performance of your Spex NanoSNAP™ spectrophotometer
  • Regular maintenance of your spectrophotometer will ensure accurate and precise results
$115.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • ConcentrationNano-hydrophobic coating maintenance kit
  • Materials of ConstructionNano-hydrophobic coating solution and swabs
  • DescriptionNanoSNAP™ Maintenance Kit, Hydrophobic Reagent and Swabs
Key Features
  • Regular maintenance of your spectrophotometer will ensure accurate and precise results
More About this Item
This Spex NanoSNAP™ maintenance kit features a nano-hydrophobic coating solution and swabs to optimize performance of your Spex NanoSNAP™ microvolume spectrophotometer. Spectrophotometer, 83057-02, is sold separately.
For the best performance, it is suggested to do annual maintenance on the Spex NanoSNAP. It includes light path confirmation and nano-coating layer recoating.