Spex SamplePrep 2010 Geno/Grinder® High-Throughput Homogenizer; 115 VAC, 50/60 Hz

Representative image only.
Spex SamplePrep Item # EW-04500-17
Accommodates full range of sample vials from 2 to 50 mL
  • Fast and efficient extraction of nucleic acids, proteins, and other molecules
  • Double-locking lid provides safe operation
  • Sound insulation and suspension system ensure quiet, vibration-free operation
  • Fully adjustable clamp allows simple front loading of vials or microplates
  • Interior LED lighting and large window improve sample visibility

PRODUCT OPTIONS

$17,920.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Min Sample Size (mL)2
  • Max Sample Size (mL)50
  • Height (in)28
  • Width (in)14
  • Depth (in)22 1/2
  • Height (cm)71.1
  • Width (cm)35.6
  • Depth (cm)57.2
  • Power (VAC)115
  • Power (Hz)50/60
  • Motor hP0.5
  • DescriptionHigh-Throughput Homogenizer; 115 VAC, 50/60 Hz

Includes

Adjustable clamp assembly and three-prong grounded plug with cord.

Key Features

  • Fast and efficient extraction of nucleic acids, proteins, and other molecules
  • Double-locking lid provides safe operation
  • Sound insulation and suspension system ensure quiet, vibration-free operation
  • Fully adjustable clamp allows simple front loading of vials or microplates
  • Interior LED lighting and large window improve sample visibility
  • Fully programmable control screen tilts for optimal viewing angle
  • Adjustable clamp speed from 500 to 1700 strokes/min
  • Clamp provides 1.25" (3.2 cm) vertical movement
  • 115 VAC, 50/60 Hz operation

More About this Item

The 2010 Geno/Grinder automated high-throughput homogenizer and cell lyser is designed for rapid cell disruption of plant and animal tissue through bead beating. This process offers fast and efficient extraction of nucleic acids, proteins, and other molecules. Samples can be processes with or without reagents in approximately 60 seconds or less.

An adjustable clamp features unique vigorous vertical grinding motion allowing increased reproducibility, superior extraction efficiency, and consistency of every sample. The clamp can accommodate a full range of sample vials from 2 to 50 mL centrifuge tubes or up to six deep-well microplates. Cryogenic accessories (sold separately) are available to allow you to homogenize temperature-sensitive samples such as proteins and RNA.

The password-protected, touch screen control panel enables you to program run time, rate, cycles, and pause time. The control panel can be tilted for better viewing. Up to 500 protocols can be easily labeled, saved, recalled, and exported via USB.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat