Spex SamplePrep 2060 Genoseal™ Semi-Automatic Heat Sealer for Microplates and Tube Racks; 115/220 VAC

Representative image only.
Spex SamplePrep Item # EW-41071-39
Designed to provide permanent or peelable seals on most types of microplates
  • Utilizes various heat seal films and foils to give either permanent or peelable seals for all common microplate material types
  • Ensure a perfect seal with fully adjustable sealing times and temperatures via the control touchscreen
  • Fully compatible with full-skirted ANSI/SLAS standard plates and PCR plates
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Plate TypeSuitable for plates with 9 mm to 54 mm height
  • Min Temperature (° C)145
  • Max Temperature (° C)200
  • Height (in)12 1/8
  • Width (in)15 5/16
  • Depth (in)8 1/16
  • Height (mm)308
  • Width (mm)389
  • Depth (mm)205
  • Power (VAC)115 / 220
  • Power (Hz)50/60
  • DescriptionGenoseal™ Semi-Automatic Heat Sealer for Microplates and Tube Racks; 115/220 VAC
  • Warranty1 Year
1 Year Warranty

Includes

41071-35 and 41071-36 plate weights, 41071-40 plate locator, and power cord.

Key Features

  • Utilizes various heat seal films and foils to give either permanent or peelable seals for all common microplate material types
  • Ensure a perfect seal with fully adjustable sealing times and temperatures via the control touchscreen
  • Fully compatible with full-skirted ANSI/SLAS standard plates and PCR plates
  • 1 year warranty

More About this Item

The Spex® SamplePrep Genoseal™ semi-automatic heat sealer provides a permanent or peelable seal for deep-well, shallow-well, low-profile deep-well, filter, and assay plates that conform to the ANSI/SLAS standard format. The Genoseal operates using 400 W of heating elements that ensure high integrity sealing of almost any type of microplate. With so much available power, sealing times for most plates are five seconds or less. Sealed plates are automatically ejected from the unit when the sealing cycle has completed. The compact footprint means that the Genoseal will fit just about anywhere—lab bench or inside a fume hood, if required.

The Genoseal utilizes various heat seal films and foils to give either permanent or peelable seals for all common microplate material types such as polypropylene, polystyrene, and polyethylene. Seal times and seal temperatures are fully adjustable on the product via the control screen.

The included plate weights keep the seal flat before sealing and help prevent the plate from sticking to the heater block. The included plate locator allows for the plate skirt to sit around the corresponding locator and support the bottom of the plate. Various plate adapters are available for a variety of different types of microplates and support each cell individually to ensure the seal is even. These plate adapters are sold separately.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat