Spex SamplePrep 2060-PL Genoseal™ Plate Locator for Skirted Plates
Representative image only.
Spex SamplePrep Item # EW-41071-40
Optimize performance of the Spex® SamplePrep Genoseal™ heat sealer
  • Choose from a wide range of accessories to enhance operation of the Genoseal™
  • Allows for the plate skirt to sit around the corresponding locator and support the bottom of the plate
SPECS & DESCRIPTION
Specifications & Description
  • Plate TypePlate Locator for Skirted Plates
  • DescriptionGenoseal™ Plate Locator for Skirted Plates
Key Features
  • Choose from a wide range of accessories to enhance operation of the Genoseal™
  • Allows for the plate skirt to sit around the corresponding locator and support the bottom of the plate
More About this Item
These replacement and optional accessories are suitable for use with the Spex® SamplePrep Genoseal™ semi-automatic heat sealer, item 41071-39. Genoseal is sold separately.
Plate weights keep the seal flat before sealing and help prevent the plate from sticking to the heater block. The plate locator allows for the plate skirt to sit around the corresponding locator and support the bottom of the plate. Various plate adapters are available for a variety of different types of microplates and support each cell individually to ensure the seal is even.