Spex SamplePrep 2060-WT-STD Genoseal™ Plate Weight for Standard and Rimless Plates, Brass
Representative image only.
Spex SamplePrep Item # EW-41071-36
Optimize performance of the Spex® SamplePrep Genoseal™ heat sealer
  • Choose from a wide range of accessories to enhance operation of the Genoseal™
  • Keeps the seal flat before sealing
  • Prevents the plate from sticking to the heater block
SPECS & DESCRIPTION
Specifications & Description
  • Plate TypePlate Weight for Standard and Rimless Plates
  • MaterialBrass
  • DescriptionGenoseal™ Plate Weight for Standard and Rimless Plates, Brass
Key Features
  • Choose from a wide range of accessories to enhance operation of the Genoseal™
  • Keeps the seal flat before sealing
  • Prevents the plate from sticking to the heater block
More About this Item
These replacement and optional accessories are suitable for use with the Spex® SamplePrep Genoseal™ semi-automatic heat sealer, item 41071-39. Genoseal is sold separately.
Plate weights keep the seal flat before sealing and help prevent the plate from sticking to the heater block. The plate locator allows for the plate skirt to sit around the corresponding locator and support the bottom of the plate. Various plate adapters are available for a variety of different types of microplates and support each cell individually to ensure the seal is even.