Spex SamplePrep Vial with Screw Cap, Polypropylene, 2 mL; 500/Pk
Representative image only.
Spex SamplePrep Item # EW-04500-29
Safely grind samples in the HG-600 Geno/Grinder® 2010 and HG-400 MiniG® homogenizers
  • Screw thread cap ensures a tight seal
  • Constructed of durable, opaque, polypropylene
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)2
  • Tube MaterialPolypropylene
  • Tube ColorWhite
  • Cap TypeScrew
  • Cap MaterialPolypropylene
  • Cap colorWhite
  • SterileNo
  • DescriptionVial with Screw Cap, Polypropylene, 2 mL; 500/Pk
Includes
Vials and screw caps.
Key Features
  • Screw thread cap ensures a tight seal
  • Constructed of durable, opaque, polypropylene
More About this Item
Cole-Parmer® vials are ideal for holding grinding balls and media. Vials are supplied with screw caps to secure and protect samples or grinding media.