Cole-Parmer Spex® 2181 Acid Washed Zirconia Grinding Beads, Low-Binding, 100 µm; 250 g Bottle
Representative image only.
Cole-Parmer Item # EW-04575-55
Treated to remove fine particles and contaminants
  • Choose from silica or zirconia grinding beads to help remove fine particles and contaminant
  • Coated to reduce nonspecific binding of nucleic acids and proteins
  • Treated to keep samples from binding to titer plate wells
  • Approximately 100 µm bead size
  • 250 g bottle
PRODUCT OPTIONS
$366.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Particle Size (micron)100
  • MaterialZirconia
  • DescriptionAcid Washed Zirconia Grinding Beads, Low-Binding, 100 µm; 250 g Bottle
Key Features
  • Choose from silica or zirconia grinding beads to help remove fine particles and contaminant
  • Coated to reduce nonspecific binding of nucleic acids and proteins
  • Treated to keep samples from binding to titer plate wells
  • Approximately 100 µm bead size
  • 250 g bottle
More About this Item
Using untreated beads which contain impurities can affect homogenization and biological processes. Acid washing silica and zirconia grinding beads help remove fine particles and contaminant for cleaner samples. Select silica beads for general grinding applications; zirconia beads have greater density and offer higher impact power. General-purpose grinding beads are suitable for most basic cell lysis applications. Choose RNase/DNase-free treated molecular biology-grade grinding beads to inactivate contaminating enzymes. Low-binding grinding beads are coated to reduce nonspecific binding of nucleic acids and proteins.