Spex SamplePrep 2191 Homogenizer Foam Holder, 48 x 5 mL Vials; 1/EA

Representative image only.
Spex SamplePrep Item # EW-04500-25
Securely hold vials in place while processing samples
  • Safely grind samples in the 2010 Geno/Grinder®
  • Accommodates up to 48 x 5 mL vials 04576-98 and 04500-36

PRODUCT OPTIONS

$51.20USD / Each
$64.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Max Tube Size (mL)5
  • Tubes Held48
  • MaterialFoam
  • DescriptionHomogenizer Foam Holder, 48 x 5 mL Vials; 1/EA

Key Features

  • Safely grind samples in the 2010 Geno/Grinder®
  • Accommodates up to 48 x 5 mL vials 04576-98 and 04500-36

More About this Item

Vial holders are suitable for use with the Spex® SamplePrep 2010 Geno/Grinder® Homogenizers. Order homogenizer and vials separately.

The foam block is ideal for use in the Geno/Grinder for shaking samples in 5 mL vials.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat