Spex SamplePrep 2210-100 Deep-Well Microplate, 96 x 1 mL, Round-Bottom Wells; 100/PK

Representative image only.
Spex SamplePrep Item # EW-04500-31
Durable polypropylene microplates resist perforation by grinding media
  • Alphanumeric marks for well identification
  • Use microplates with sealing mats to prevent spills and well-to-well contamination
  • Round-bottom well has a working volume of 0.7 mL
  • Pack of 100

PRODUCT OPTIONS

$732.80USD / Pkg of 100
$916.00USD / Pkg of 100

SPECS & DESCRIPTION

Specifications & Description

  • Number of Wells96
  • Well Volume (mL)1
  • Max Working Well Volume (mL)0.7
  • Well ShapeRound
  • Lid (Included)No, order sealing mat 04500-33 or 04500-34
  • MaterialPolypropylene
  • DescriptionDeep-Well Microplate, 96 x 1 mL, Round-Bottom Wells; 100/PK

Key Features

  • Alphanumeric marks for well identification
  • Use microplates with sealing mats to prevent spills and well-to-well contamination
  • Round-bottom well has a working volume of 0.7 mL
  • Pack of 100

More About this Item

Ideal for use with Spex® SamplePrep homogenizers, these deep-well plates serve many application purposes, such as tissue homogenization and cell lysis.

Load each well with the sample, solution (if desired), and grinding media then seal with a sealing mat (04500-33 or -34) to prevent spills and cross contamination. Place up to six microplates in the 2010 Geno/Grinder 04500-17 or 04576-84 for high-throughput tissue homogenization and cell lysis.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat