Spex SamplePrep 2231 Sealing Mat for Square 24-Well Microlate; 1/EA
Representative image only.
Spex SamplePrep Item # EW-04575-69
Tightly seals microplate wells to prevents spills and well-to-well contamination
  • Constructed of rugged silicone rubber
  • May be sterilized and reused
  • Use with 24-well microplates with square wells (04575-68 and 04576-94)
  • 1 each
  • 1 year warranty
PRODUCT OPTIONS
$9.60USD / Each
$12.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialSilicone
  • Compatible WithMicroplates 04575-68 and 04576-94
  • DescriptionSealing Mat for Square 24-Well Microlate; 1/EA
  • Warranty1 year
1 Year Warranty
Key Features
  • Constructed of rugged silicone rubber
  • May be sterilized and reused
  • Use with 24-well microplates with square wells (04575-68 and 04576-94)
  • 1 each
  • 1 year warranty
More About this Item
Use these flexible deep-well sealing mats to reduce maximum well volume. 