Spex SamplePrep 2665 Homogenizer Cryo-Block, Aluminum, 96 x 0.6 mL Tubes; 2/PK
Representative image only.
Spex SamplePrep Item # EW-04575-86
Securely hold vials in place while processing samples
  • Safely grind samples in the 2010 Geno/Grinder®
  • Accepts 0.6 mL microcentrifuge or PCR tubes
  • Enables high-throughput cryogenic grinding
  • Increases yields by grinding samples at sub-zero temperatures
  • Each pair holds up to 196 tubes for rapid and efficient sample preparation
PRODUCT OPTIONS
$416.80USD / Pkg of 2
$521.00USD / Pkg of 2
SPECS & DESCRIPTION
Specifications & Description
  • Max Tube Size (mL)0.6
  • Tubes Held96
  • MaterialAluminum
  • DescriptionHomogenizer Cryo-Block, Aluminum, 96 x 0.6 mL Microcentrifuge or PCR Tubes; 2/PK
  • Warranty1 year
1 Year Warranty
Key Features
  • Safely grind samples in the 2010 Geno/Grinder®
  • Accepts 0.6 mL microcentrifuge or PCR tubes
  • Enables high-throughput cryogenic grinding
  • Increases yields by grinding samples at sub-zero temperatures
  • Each pair holds up to 196 tubes for rapid and efficient sample preparation
  • 1 year warranty
More About this Item
Vial holders are suitable for use with the Spex® SamplePrep 2010 Geno/Grinder® Homogenizers. Order homogenizer and vials separately.
Each aluminum cryo-block holds 96 standard (0.6 mL) microcentrifuge or PCR tubes. Chill cryo-blocks in the cryo-station 04575-81 to maintains samples at liquid nitrogen temperatures during tissue homogenization and cell lysis. The blocks preserve pesticides, RNA, and proteins for heat sensitive extraction methods. Use two at a time in the clamp assembly 04605-08 to keep it in balance.