Cole-Parmer Spex® 31688 Friction Disk, Rubber
Representative image only.
Cole-Parmer Item # EW-04576-82
Optimize performance of your Shatterbox® Verastile Ring and Puck Mill
  • Choose from a wide range of accessories and replacement parts to ensure safe operation
  • Improves grip of Shatterbox® mill clamp
PRODUCT OPTIONS
$18.90USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialRubber
  • Compatible WithShatterBox® Ring and Puck Mill
  • DescriptionFriction Disk, Rubber
Key Features
  • Choose from a wide range of accessories and replacement parts to ensure safe operation
  • Improves grip of Shatterbox® mill clamp
More About this Item
Suitable for use with the Cole-Parmer® SM-200 ShatterBox® Ring and Puck Mills, 04578-25 and 04578-26. Mills sold separately.
Place under grinding container to reduce wear on capsule retainer and control operating noise.